Streptavidin agarose, Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Pierce™ Streptavidin agarose is a standard-capacity crosslinked 6% beaded agarose resin of immobilised recombinant streptavidin protein for use in a variety of biotin-binding affinity purification methods.
- Purified recombinant streptavidin protein (53 kDa, near-neutral pI); tetrameric with four biotin-binding site per molecule
- Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
- Streptavidin is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
- Standard capacity beads with a normal load of immobilised streptavidin, providing a binding capacity of 1 to 3 mg of biotinylated BSA per ml of resin
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together