Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amberlite® XAD4
Amberlite® XAD4 ion exchange resin
Amberlite® XAD4 ion exchange resin
:
Thermo Fisher Scientific
Amberlite® XAD4 ion exchange resin
:
Thermo Fisher Scientific
:
37380-42-0
:
9003-69-4
:
MFCD00132704
:
14,382
:
Ambient
Environmentally Preferable:
msds_ALFAL14142
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Reag. USP, ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
VWR®, Qualitative Filter Papers
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals