TRIS-glycine native sample buffer (4X), pH 8,6
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical Form: Liquid
Appearance: Colourless to yellow
MDL No.: MFCD00236360
Applications: For preparation of protein samples for native gel electrophoresis
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together