Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, Methyl 3,4-dihydro-3-oxo-2H-1,4-benzothiazine-6-carboxylate
CAS Number:188614-01-9
:
Formula:C₁₀H₉NO₃S
MDL Number:MFCD00449104
Molecular Weight:223.25 g/mol
Melting Pt:97…99 °C
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together