Polyethylene glycol methyl ether methacrylate, M.W. 5,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Polyethylene glycol monomethyl ether Methacrylate, mPEG Methacrylate, Methoxy PEG Methacrylate
CAS Number:26915-72-0
:
Formula:CH₃(OCH₂CH₂)nOCOC(=CH₂)CH₃
MDL Number:MFCD00241432
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together