Polydimethylsiloxane emulsion
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
CAS Number:63148-62-9
:9016-00-6
Formula:(CH₃)₃SiO(Si(CH₃)₂O)n
MDL Number:MFCD00084411
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together