Polyethyleneimine, M.W. M.W. 50,000 - 100,000 30% (w/w) in aqueous solution, branched
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
CAS Number:9002-98-6
:
Formula:(CH2CH2NH)n
MDL Number:MFCD00084427
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together