Serum separator tubes, SST™, BD Vacutainer®
Catalog # BDAM367985
Supplier: BD Medical
Specifications
- Closure type:Standard closure
- Ø:16 mm
- Volume:10 ml
- Description:Blood Collection tube, SST™
- Length:125 mm
- Material:Plastic
- Environmentally Preferable:
- Cap colour:Red/grey
- Pk:1.000
- Additive:Double polymer gel cot activator
Specifications
About this item
BD Vacutainer® SST™ Tubes contain spray-coated silica to aid in clotting and a polymer gel for serum separation. Samples processed in these tubes are used for serum determinations in chemistry, blood donor screening and infectious disease testing.* BD Vacutainer® SST™ tubes provide an efficient means of serum sample preparation and help to improve laboratory workflow.