Magnetic beads, Sera-Mag SpeedBeads Amine-blocked particles
Supplier: Cytiva
Certificates
About this item
Sera-Mag SpeedBeads Amine-Blocked Particles reduce undesired protein adsorption.
- Non-surfactant, non-protein blocking reagent provides very low non-specific binding of proteins from the sample matrix
- Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
- Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
- Uniform, nominal 1 μm diameter provides excellent lot-to-lot reproducibility
Sera-Mag SpeedBeads Amine-Blocked Particles combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics. Typical applications include molecular biology, nucleic acid isolation, research, and clinical diagnostic immunoassays.
Related Information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together