Protective and regenerating hand cream, Stokoderm® Aqua PURE
Aqua PURE is a barrier cream (W/O/W emulsion) that protects the skin when working with aqueous or water-miscible industrial substances and performing wet work. It has a repairing effect and stabilises, regenerates, and strengthens the skin barrier.
- Contains bisabolol and witch hazel to ensure a high level of protection and care
- Compatible with alcoholic hand disinfectants; does not impair the disinfection performance, in accordance with prEN 12054
- Free from preservatives, perfume, colourants, and silicone
Suitable for use in industry, hospitals, medical facilities and laboratories, the food industry, and catering. Quick absorption and ease of application means it can also be used in areas where good hygiene is essential. Suitable for use on hands, face and body. Also suitable for use with gloves.
: HACCP
Frequently Bought Together