Order Entry
Slovenia
ContactUsLinkComponent
Anti-FEM 1B Rabbit Polyclonal Antibody
Anti-FEM 1B Rabbit Polyclonal Antibody
Catalog # BOSSBS-7113R
Supplier:  Bioss
undefined
Anti-FEM 1B Rabbit Polyclonal Antibody
Catalog # BOSSBS-7113R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    fem-1 homolog b (C. elegans)
  • Antigen symbol:
    FEM 1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Gene ID:
    10116
  • Antigen synonyms:
    Fem-1-like in apoptotic pathway protein alpha|Protein fem-1 homolog B|F1A-ALPHA|Fem-1-like death receptor-binding protein alpha|KIAA0396|F1AA|FEM1b|FEM1-beta
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    550-600/627
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit. Involved in apoptosis by acting as a death receptor-associated protein that mediates apoptosis. Also involved in glucose homeostasis in pancreatic islet. Functions as an adapter/mediator in replication stress-induced signaling that leads to the activation of CHEK1.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FEM 1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat