Order Entry
Slovenia
ContactUsLinkComponent
Anti-E2F2 Antibody Pair
Anti-E2F2 Antibody Pair
Catalog # ABNODI0571
Supplier:  Abnova
Anti-E2F2 Antibody Pair
Catalog # ABNODI0571
Supplier:  Abnova

Specifications

  • Antigen symbol:
    E2F2
  • Host:
    Multiple
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Gene ID:
    1870|1869
  • Antigen synonyms:
    E2F-2
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the E2F2 protein, and the other against the E2F1 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. E2F2 Rabbit purified Polyclonal Antibody (20 µg) 2. E2F1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: E2F2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: