Bottle-top dispensers, Dispensette® S Trace Analysis
: Brand
Analogue-adjustable.
- A field-tested cleaning process before use in trace analysis is described in the operating manual
- If contamination of the bottle contents must be avoided, we recommend using a device without a recirculation valve
- 45 mm standard thread plus the included adapters fit most common lab bottles
- Valve block can be rotated 360° so that the bottle label always faces the user for safety
- Telescopic filling tube adjusts to different bottle sizes
- Easy to take apart when replacing the dispensing cartridge
The Dispensette® S Trace Analysis provides outstanding performance for precise volume dispensing of high purity media for trace analysis. Also suitable for HF with platinum-iridium valve spring. Plastics in contact with media consist of high purity materials such as PTFE, ETFE, FEP and PFA. The purest sapphire is used for replaceable valves. Depending on the application, platinum-iridium or tantalum are available as spring materials.
: Dispensette® S Trace Analysis bottle-top dispenser supplied with DE-M marking, performance certificate, telescopic filling tube, recirculation tube (optional), mounting tool and bottle adapters GL 28/S 28 (ETFE), GL 32 (ETFE) and S 40 (PTFE).
- Catalog No:
- 613-5357
- 613-5356
- 613-5355
- 613-5354
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
Frequently Bought Together