Anti-EFEMP2 Mouse Monoclonal Antibody [clone: 2F12]
ABNOH00030008-M29
: Abnova
- Antibody type:Primary
- Antigen name:EGF containing fibulin-like extracellular matrix protein 2
- Antigen symbol:EFEMP2
- Clonality:Monoclonal
- Clone:2F12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:30008
- Antigen synonyms:ARCL1B|MBP1|FBLN4|UPH1
- Amino acid number:26 to 443
- Storage buffer:1x PBS, pH 7,4
- Sequence:SPQDSEEPDSYTECTDGYEWDPDSQHCRDVNECLTIPEACKGEMKCINHYGGYLCLPRSAAVINDLHGEGPPPPVPPAQHPNPCPPGYEPDDQDSCVDVDECAQALHDCRPSQDCHNLPGSYQCTCPDGYRKIGPECVDIDECRYRYCQHRCVNLPGSFRCQCEPGFQLGPNNRSCVDVNECDMGAPCEQRCFNSYGTFLCRCHQGYELHRDGFSCSDIDECSYSSYLCQYRCVNEPGRFSCHCPQGYQLLATRLCQDIDECESGAHQCSEAQTCVNFHGGYRCVDTNRCVEPYIQVSENRCLCPASNPLCREQPSSIVHRYMTITSERSVPADVFQIQATSVYPGAYNAFQIRAGNSQGDFYIRQINNVSAMLVLARPVTGPREYVLDLEMVTMNSLMSYRASSVLRLTVFVGAYTF
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:EFEMP2 (AAH10456.1, 26 a.a. ~ 443 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against a full-length recombinant EFEMP2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: EFEMP2
Clonality: Monoclonal
Clone: 2F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: