Order Entry
Slovenia
ContactUsLinkComponent
Anti-CUX2 Mouse Monoclonal Antibody [clone: 2H8]
Anti-CUX2 Mouse Monoclonal Antibody [clone: 2H8]
Catalog # ABNOH00023316-M03
Supplier:  Abnova
Anti-CUX2 Mouse Monoclonal Antibody [clone: 2H8]
Catalog # ABNOH00023316-M03
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cut-like homeobox 2
  • Antigen symbol:
    CUX2
  • Clonality:
    Monoclonal
  • Clone:
    2H8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Liquid
  • Gene ID:
    23316
  • Antigen synonyms:
    CUTL2|CDP2
  • Amino acid number:
    121 to 220
  • Storage buffer:
    In 1x PBS, pH 7,4
  • Sequence:
    FDPSGQPRRDLHTSWKRNPELLSPKEQREGTSPAGPTLTEGSRLPGIPGKALLTETLLQRNEAEKQKGLQEVQITLAARLGEAEEKIKVLHSALKATQAE
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    CUX2 (NP_056082.1, 121 a.a. ~ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against a partial recombinant CUX2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CUX2
Clonality: Monoclonal
Clone: 2H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human