Anti-SMC2 Mouse Monoclonal Antibody [clone: E1M]
ABNOMAB1840
: Abnova
- Antibody type:Primary
- Antigen name:Structural Maintenance of Chromosomes 2
- Antigen symbol:Structural maintenance of chromosomes protein 2
- Clonality:Monoclonal
- Clone:E1M
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:10592
- Antigen synonyms:SMC2L1|structural maintenance of chromosomes 2|CAP-E|SMC2|human chromosome-associated protein E|structural maintenance of chromosomes 2-like 1|CAPE|hCAP-E
- Amino acid number:523 to 768
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 523-768 of human SMC2.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against partial recombinant SMC2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: SMC2
Clonality: Monoclonal
Clone: E1M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human