Anti-GAL4 Mouse Monoclonal Antibody [clone: 5C8]
ABNOMAB1883
: Abnova
- Antibody type:Primary
- Antigen name:Galectin 4
- Antigen symbol:GAL4
- Clonality:Monoclonal
- Clone:5C8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Yeast
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:855828
- Antigen synonyms:galectin 4|L-36 lactose-binding protein|antigen NY-CO-27|lactose-binding lectin 4|gal-4|galectin-4|GAL4|L36LBP
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length yeast GAL4.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against full length recombinant GAL4.
Type: Primary
Antigen: GAL4
Clonality: Monoclonal
Clone: 5C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Yeast