Anti-YFP Mouse Monoclonal Antibody [clone: 64a59]
ABNOMAB8759
: Abnova
- Antibody type:Primary
- Antigen symbol:YFP
- Clonality:Monoclonal
- Clone:64a59
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Jellyfish
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with carrier KLH) corresponding to N-terminus amino acids of Aequorea victoria YFP.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of YFP.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: YFP
Clonality: Monoclonal
Clone: 64a59
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Jellyfishes