Anti-STMN1 Mouse Monoclonal Antibody [clone: 678r]
Catalog # ABNOMAB8841
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Stathmin 1
- Antigen symbol:STMN1
- Clonality:Monoclonal
- Clone:678r
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:3925
- Antigen synonyms:Op18|SMN|C1orf215|PP19|LAP18|OP18|Stathmin|Lag|PR22|Leukemia-associated phosphoprotein p18|PP17
- Storage buffer:citrate-Tris-HCl, pH7.0 (0,02% Procl300)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length of human STMN1.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant STMN1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: STMN1
Clonality: Monoclonal
Clone: 678r
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human