Anti-R3hdm1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB15706
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:R3H domain containing 1
- Antigen symbol:R3HDM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:226412
- Antigen synonyms:R3hdm
- Storage buffer:PBS (50% glycerol, 0,02% sodium azide)
- Sequence:YPLLGQPLQYNPPTLLHGHIPHQQGQSGSRHGNRGRRQAKKAASTDLGAGEAVVGKVLEITELPDGITRVEAEKLFGELFKIGAKIRWLRDPQSQPQLRRHALCCGSGDNTVNPEHSKPSDLASTYTVLATFPSISAAQSALKKQIHSVNKFKLRMSKKHYDFHILERASSQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to 172 mouse R3hdm1.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant R3hdm1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: R3hdm1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse