Anti-ARHGEF17 Rabbit Polyclonal Antibody
ABNOPAB15715
: Abnova
- Antibody type:Primary
- Antigen name:Rho Guanine Nucleotide Exchange Factor 17
- Antigen symbol:ARHGEF17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:207212
- Antigen synonyms:TEM4|P164RHOGEF|Tumor Endothelial Marker 4|KIAA0337|P164-RhoGEF
- Storage buffer:PBS (50% glycerol, 0,02% sodium azide)
- Sequence:MLAGSDAIIRQHKAACLRITALLVCAELLWVGTSAGVVLTIPTSPSTVSCPRAPLSPAGLCQGHTGHVRFLAAVQLPEGFNLLCSTPPPPPDTGPEKLPSLDHRDSPRRRGPTSARPKMLVISGGDGSEDFRLSSGGGGSSETVGRDDSTNHLLLWRV
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to 158 mouse Arhgef17.
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against partial recombinant Arhgef17.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: ARHGEF17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse