Anti-ANLN Goat Polyclonal Antibody
Catalog # ABNOPAB6482
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Anillin, Actin Binding Protein
- Antigen symbol:ANLN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:54443
- Antigen synonyms:scra|FSGS8|Scraps
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:WQPDACYKPIGKP
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human ANLN.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of ANLN.
Recommended Dilutions: ELISA: 1:64000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 3-10 ?g/ml
Type: Primary
Antigen: ANLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human