Anti-GC Goat Polyclonal Antibody
Catalog # ABNOPAB17040
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Group-specific component
- Antigen symbol:GC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:2638
- Antigen synonyms:VDBP|GRD3|DBP/GC|DBP|VDBG|HEL-S-51
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:ERGRDYEKNKVCK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human GC.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of GC.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 0.5-3 ?g/ml
Type: Primary
Antigen: GC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human