Anti-VTI1B Goat Polyclonal Antibody
ABNOPAB7012
: Abnova
- Antibody type:Primary
- Antigen name:Vesicle Transport Through Interaction with t-SNAREs 1B
- Antigen symbol:VTI1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:10490
- Antigen synonyms:vesicle-associated soluble NSF attachment protein receptor|VTI1B|v-SNARE|vesicle transport through interaction with t-SNAREs homolog 1B (yeast) |VTI1|VTI1L|vesicle transport through interaction with t-SNAREs 1B|VTI2 |HGNC:17793
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-RDFDEKQQEANET
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human VTI1B.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of VTI1B.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: VTI1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human