Anti-TMBIM6 Mouse Monoclonal Antibody (Unconjugated) [clone: 20F430]
ROCK200-301-H60
- Antibody type:Primary
- Antigen name:transmembrane BAX inhibitor motif containing 6
- Antigen symbol:TMBIM6
- Clonality:Monoclonal
- Clone:20F430
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:No
- Gene ID:7009
- Antigen synonyms:BAXI1|BI-1|TEGT
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,05% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Bi-1 Antibody was produced in mice prepared by repeated immunizations of a mixture of three synthetic peptides corresponding to the N-terminus, internal and C-terminus of human Bi-1 protein.
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Anti-Bi-1 antibody is suitable for use in IHC (p). Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 5-10 µg/ml
Type: Primary
Antigen: TMBIM6
Clonality: Monoclonal
Clone: 20F430
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human