Anti-LAMP2 Rat Monoclonal Antibody (Unconjugated) [clone: GL2A7]
ROCK200-501-G10
- Antibody type:Primary
- Antigen name:lysosomal-associated membrane protein 2
- Antigen symbol:LAMP2
- Clonality:Monoclonal
- Clone:GL2A7
- Conjugation:Unconjugated
- Host:Rat
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:16784
- Antigen synonyms:LGP-B|Lamp-2c|Lamp-2a|Lamp-2|Mac3|Lamp-2b|CD107b|Lamp II
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:LAMP2 Antibody was produced in rat by repeated immunizations with purified preparation of mouse liver lysosomal membranes.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Anti-LAMP2 Antibody is recommended for use in IF microscopy and IP. The antibody will label the presumptive lysosomes and late endosomes in cells that have been permeabilized with saponin.
Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone: GL2A7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rabbit, Mouse