Anti-Visual System Homeobox Protein 2 Rabbit Polyclonal Antibody
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Visual System Homeobox Protein 2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat,
- Catalog No:
- USBI142805-100
- USBI142805
- Antigen symbol:
- VSX2
- VSX2
- Antigen name:
- Visual System Homeobox Protein 2
- Visual System Homeobox Protein 2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Rat
- Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- VSX2 (Pro2~His135); MGHHHHHHSGSEF-PVDFNGYWK MLSNENFEEY LRALDVNVAL RKIANLLKPD KEIVQDGDHM IIRTLSTFRN YIMDFQVGKE FEEDLTGIDD RKCMTTVSWD GDKLQCVQKG EKEGRGWTQW IEGDELHLEM RAEGVTCKQV FKKVH
- VSX2 (Pro2~His135); MGHHHHHHSGSEF-PVDFNGYWK MLSNENFEEY LRALDVNVAL RKIANLLKPD KEIVQDGDHM IIRTLSTFRN YIMDFQVGKE FEEDLTGIDD RKCMTTVSWD GDKLQCVQKG EKEGRGWTQW IEGDELHLEM RAEGVTCKQV FKKVH
- Amino acid number:
- 2 to 135
- 2 to 135
- Sequence:
- MGHHHHHHSGSEFPVDFNGYWKMLSNENFEEYLRALDVNVRKIANLLKPDKEIVQDGDHMIIRTLSTFRNYIMDFQVGKEFEEDLTGIDDRKCMTTVSWDGDKLQCVQKGEKEGRGWTQWIEGDELHLEMRAEGVTCKQVFKKVH
- MGHHHHHHSGSEFPVDFNGYWKMLSNENFEEYLRALDVNVRKIANLLKPDKEIVQDGDHMIIRTLSTFRNYIMDFQVGKEFEEDLTGIDDRKCMTTVSWDGDKLQCVQKGEKEGRGWTQWIEGDELHLEMRAEGVTCKQVFKKVH
- Form:
- liquid
- liquid
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- 200 ug/ml
- 200 ug/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- Storage temperature:
- –20 °C
- –20 °C
Frequently Bought Together