Anti-KRT23 Mouse Monoclonal Antibody [clone: 13L5]
Recommended dilution: Western Blot: 1:500-1:3000
Type: Primary
Antigen: KRT23
Clonality: Monoclonal
Clone: 13L5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
- Catalog No:
- USBI138682
- USBI138682-100
- Antigen symbol:
- KRT23
- KRT23
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 13L5
- 13L5
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant protein corresponding to aa271-422 from human KRT23 expressed in E. coli.
- Recombinant protein corresponding to aa271-422 from human KRT23 expressed in E. coli.
- Amino acid number:
- 271 to 422
- 271 to 422
- Form:
- liquid
- liquid
- Purification:
- purified
- purified
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,4, 0,1% sodium azide.
- PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:
- –20 °C
- –20 °C
Frequently Bought Together