Anti-Talin 2 Mouse Monoclonal Antibody [clone: 53.8]
USBIT0751-35G-100
- Antibody type:Primary
- Antigen symbol:Talin 2
- Clonality:Monoclonal
- Clone:53.8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Talin-2
- Purification:purified by Protein G chromatography
- Size:100 μg
- Pk:100 µG
Recommended dilution: Immunochemistry: 1:25-1:100
Type: Primary
Antigen: Talin 2
Clonality: Monoclonal
Clone: 53.8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human,