Anti-AGP-A Rabbit Polyclonal Antibody
Catalog # ORIGTA327179
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:alpha-1-Acid Glycoprotein
- Antigen symbol:AGP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Orosomucoid 1|ORM1
- Storage buffer:Store at -20C or -80C. Avoid freeze / thaw cycles. Buffer: PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Recombinant protein of human ORM1
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: AGP-A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse