Anti-ANG-2 Rabbit Polyclonal Antibody
Catalog # ORIGTA319449
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Angiopoietin 2
- Antigen symbol:ANGPT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:near N-terminus
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:ANG2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide, corresponding to a region near the N-terminus of mouse angiopoietin-2 protein, conjugated to KLH using maleimide.
- Size:200 μl
- Pk:200 µl
Specifications
About this item
Recommended Dilutions: ELISA: 1:5000-1:25000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: ANG-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse