- Antibody type:Primary
- Antigen name:Y box binding protein 1
- Antigen symbol:YBX1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:4904
- Antigen synonyms:EFI-A|YB1|DBPB|CBF-A|NSEP-1|CSDA2|NSEP1|MDR-NF1|BP-8|CSDB|YB-1
- UniProtKB:P67809
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human YB1 (293-320aa ENPKPQDGKETKAADPPAENSSAPEAEQ), identical to the related mouse and rat sequences.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Rabbit IgG polyclonal antibody for Nuclease-sensitive element-binding protein 1(YBX1) detection. Tested with WB, IHC-P, IHC-F in Human;Mouse;Rat.
YBX1(Y box binding protein 1), commonly referred to as "YB-1" by researchers, is a human protein. YB1 binding has an absolute requirement for the CCAAT box and relative specificity for the Y box. It has a molecular mass of 35,414 and contains 18% basic residues and putative nuclear localization signals. The YBX1 gene is mapped on 1p34.2. Ybx1 was highly expressed in mouse erythroid myeloid lymphoid clone-1 (EML), a hematopoietic precursor cell line, but that it was downregulated in myeloid progenitors and in Gmcsf-treated EML cells during myeloid differentiation. Ybx1 was expressed at high levels in myeloid leukemic cells at different developmental stages. Knockdown of YBX1 in a human leukemic cell line inhibited proliferation ability, induced apoptosis, and induced megakaryocytic differentiation in response to arsenic trioxide treatment. YBX1 is downregulated during myeloid differentiation and aberrant YBX1 expression in leukemic cells may contribute to leukemia development by blocking differentiation.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for YB1 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: YES1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human