Anti-PLXNA2 Rat Monoclonal Antibody [clone: 11D24]
USBIP4271-14B
- Antibody type:Primary
- Antigen name:Plexin A2
- Antigen symbol:PLXNA2
- Clonality:Monoclonal
- Clone:11D24
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBSwith 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 2?g/ml
Type: Primary
Antigen: PLXNA2 (plexin A2)
Clonality: monoclonal
Clone: 11D24
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse