Anti-Inducible Nitric Oxide Synthase Rabbit Polyclonal Antibody
USBII0800-07G
- Antibody type:Primary
- Antigen name:Inducible nitric oxide synthase
- Antigen symbol:INOS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:17 to 31
- Storage buffer:PBS containing 1mg/ml BSA and 0,05% sodium azide
- Storage temperature:Cool
- Concentration:~4 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:200 µl
- Pk:200 µl
Recommended dilution: Immunochemistry: 1:50; Western Blot: 1:200
Type: Primary
Antigen: Inducible Nitric Oxide Synthase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat