Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-EXT2 Goat polyclonal antibody
  USBIE9008-72
Anti-EXT2 Goat polyclonal antibody
  USBIE9008-72

 

  • Antibody type:
    Primary
  • Antigen name:
    Exostosin 2
  • Antigen symbol:
    EXT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
  • Sequence:
    C-PLKVVEHRADPVL
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: ELISA: 1:8000

Type: Primary
Antigen: EXT2 (exostosin glycosyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat