Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-CARD8 Mouse Polyclonal Antibody
  USBI124342
Anti-CARD8 Mouse Polyclonal Antibody
  USBI124342

 

  • Antibody type:
    Primary
  • Antigen name:
    caspase recruitment domain family, member 8
  • Antigen symbol:
    CARD8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    MEKKECPEKSSSSEEELPRRDSGSSRNIDASKLIRLQGSRKLLVDNSIRELQYTKTGIFFQAEACVTNDTVYRELPCVSETLCDISHFFQEDDETEAEPLLFRAVPECQLSGGDIPSVSEEQESSEGQDSGDICSEENQIVSSYASKVCFEIEEDYKNRQFLGPEGNVDVELIDKSTNRYSVWFPTAGWYLWSATGLGFLVRDEVTVTIAFGSWSQHLALDLQHHEQWLVGGPLFDVTAEPEEAVAEIHLPHSISLQAGEVDVSWFLVAHFKNEGMVLEHPARVEPFYAVLESPSFSLMGILLRIASGTRLSIPITSNTLIYYHPHPEDIKFHLYLVPSDALLTKAIDDEEDRFHGVRLQTSPPMEPLNFGSSYIVSNSANLKVMPKWISSL
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: CARD8 (caspase recruitment domain family, member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human