Anti-Glutathione S Transferase Mouse Monoclonal Antibody [clone: 9AT106]
USBI031005
- Antibody type:Tag
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Monoclonal
- Clone:9AT106
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Tag
Antigen: Glutathione S Transferase
Clonality: Monoclonal
Clone: 9AT106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: