Anti-Mouse IL-1 beta Rabbit Polyclonal Antibody
ROCK210-401-319
- Antibody type:Primary
- Antigen name:Interleukin 1 beta
- Antigen symbol:IL1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Gene ID:16176
- Antigen synonyms:Il-1b|IL-1beta
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:1 mg
Type: Primary
Antigen: Mouse IL-1ß
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse