Anti-ANK3 Mouse Monoclonal Antibody
Catalog # QEDB56519
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:ankyrin 3, Node Of Ranvier (ankyrin G)
- Antigen symbol:ANK3
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ANK3 (ankyrin 3, node of Ranvier (ankyrin G))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat