Anti-6X His tag Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AD1.1.10]
Catalog # NOVUNB100-64151
Supplier: Novus Biologicals
Specifications
- Antibody type:Tag
- Antigen name:6X HIS Affinity Tag
- Antigen symbol:6xHis
- Clonality:Monoclonal
- Clone:AD1.1.10
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Multiple
- Environmentally Preferable:
- Antigen synonyms:Histidine Epitope Tag|His Tag|Histidine Tag
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pan