Anti-CASP2 Mouse monoclonal antibody unconjugated [clone: 18E809.3]
Catalog # NOVUNB100-56686
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Caspase 2
- Antigen symbol:CASP2
- Clonality:Monoclonal
- Clone:18E809.3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:835
- Antigen synonyms:regulatory subunit 57|CASP-2|ICH1|ICH-1L/1S|protein phosphatase 1|neural precursor cell expressed developmentally down-regulated protein 2|NEDD-2|protease ICH-1|caspase-2|ICH-1L|NEDD2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µL
- Pk:0,2 mL
Specifications
About this item
Type: Primary
Antigen: CASP2 (caspase 2, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 18E809.3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human