Anti-PC1/3 Rabbit Polyclonal Antibody
Catalog # BSBTPA2084
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Proprotein Convertase Subtilisin/Kexin Type 1
- Antigen symbol:PC1/3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:SPC3|PC3|BMIQ12|PC1|NEC1
- Amino acid number:726 - 740
- Sequence:DVFYNTKPYKHRDDR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human PC1/3 (726-740aa DVFYNTKPYKHRDDR), identical to the related mouse and rat sequences
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Neuroendocrine convertase 1(PCSK1) detection. Tested with WB, IHC-P in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: PC1/3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat