Anti-DKK1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1462
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Dickopf WNT Signaling Pathway Inhibitor 1
- Antigen symbol:DKK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:SK|DKK-1
- Amino acid number:68 - 83
- Sequence:YPGGNKYQTIDNYQPY
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human DKK1 (68-83aa YPGGNKYQTIDNYQPY), different from the related mouse and rat sequences by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Dickkopf-related protein 1(DKK1) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: DKK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat