Anti-PIGH Rabbit Polyclonal Antibody
BIRBORB96604-50
: Biorbyt
- Antibody type:Primary
- Antigen name:phosphatidylinositol glycan anchor biosynthesis, class H
- Antigen symbol:PIGH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Storage buffer:PBS, pH 7,4, 150 mM sodium chloride, 0,02% sodium azide, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide from human PIGH
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
ELISA: 1:40000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: PIGH (phosphatidylinositol glycan anchor biosynthesis, class H)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: