Anti-PPM1A Mouse Monoclonal Antibody
Catalog # BIRBORB95062-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:protein phosphatase, Mg2+/Mn2+ dependent, 1A
- Antigen symbol:PPM1A
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: PPM1A (protein phosphatase, Mg2+/Mn2+ dependent, 1A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human