Anti-LMP1 Rabbit Polyclonal Antibody
BIRBORB87381-50
: Biorbyt
- Antibody type:Primary
- Antigen symbol:LMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 5 ?g/ml; Western Blot: 2-4 ?g/ml
Type: Primary
Antigen: LMP1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat