Anti-UGGT2 Rabbit Polyclonal Antibody
BIRBORB77645-100
: Biorbyt
- Antibody type:Primary
- Antigen name:UDP-glucose glycoprotein glucosyltransferase 2
- Antigen symbol:UGGT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Recombinant protein
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:20000-1:80000; Western Blot: 1:500-1:1000
Type: Primary
Antigen: UGGT2 (UDP-glucose glycoprotein glucosyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human