Anti-NPSR1 Rabbit Polyclonal Antibody
Catalog # BIRBORB77451-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Neuropeptide S Receptor 1
- Antigen symbol:NPSR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Peptide
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:40000-1:80000; Western Blot: 1:200-1:1000
Type: Primary
Antigen: NPSR1 (neuropeptide S receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human