Anti-SLC29A4 Mouse Monoclonal Antibody [clone: 6B6]
Catalog # ABNOH00222962-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 29 Member A4
- Antigen symbol:Solute carrier family 29, member 4
- Clonality:Monoclonal
- Clone:6B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:222962
- Antigen synonyms:ENT4|PSEC0113|Solute carrier family 29 member 4|hENT4|Plasma membrane monoamine transporter|Equilibrative nucleoside transporter 4|PMAT|SLC29A4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SLC29A4
Clonality: Monoclonal
Clone: 6B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat