Anti-UBE2Q2 Mouse Monoclonal Antibody [clone: 2H3]
Catalog # ABNOH00092912-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating enzyme E2R 2
- Antigen symbol:UBE2Q2
- Clonality:Monoclonal
- Clone:2H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:92912
- Antigen synonyms:CDC34B|UBC3B|E2-CDC34B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: UBE2Q2
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human